ENA040899063 Enamine

{(2-furylmethyl)[(4-methylphenyl)sulfonyl]amino}acetic acid

  • MDL number MFCD06655139
  • Cas Number : 854036-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10765

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10765
Availability details for
Quantity
  • Availability
  • Add to Cart