ENA040899068 Enamine

3-{[(2-bromophenyl)sulfonyl]amino}propanoic acid

  • MDL number MFCD06353536
  • Cas Number : 781626-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10781

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10781
Availability details for
Quantity
  • Availability
  • Add to Cart