ENA040899070 Enamine

3-amino-4-(diethylamino)benzenesulfonamide

  • MDL number MFCD06353537
  • Cas Number : 41893-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102...104
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10785

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10785
Availability details for
Quantity
  • Availability
  • Add to Cart