ENA040899075 Enamine

7-chloro-3-[(oxolan-2-yl)methyl]-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD06655149
  • Cas Number : 851169-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-10794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10794
Availability details for
Quantity
  • Availability
  • Add to Cart