ENA040899077 Enamine

2-{2',5'-dioxo-2,3-dihydrospiro[1-benzopyran-4,4'-imidazolidin]-1'-yl}acetic acid

  • MDL number MFCD06655150
  • Cas Number : 851170-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99-100
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10797
Availability details for
Quantity
  • Availability
  • Add to Cart