ENA040899078 Enamine

2-fluoro-5-sulfamoylbenzoic acid

  • MDL number MFCD06355945
  • Cas Number : 112887-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-193
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10800

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10800
Availability details for
Quantity
  • Availability
  • Add to Cart