ENA040899083 Enamine

4-chloro-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD06655155
  • Cas Number : 23967-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180...182
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10812

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-10812
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart