ENA040899089 Enamine

2-chloro-N-(2,2,2-trifluoroethyl)propanamide

  • MDL number MFCD06655160
  • Cas Number : 139126-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-43
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10824

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10824
Availability details for
Quantity
  • Availability
  • Add to Cart