ENA040899090 Enamine

4-(2,5-diethylphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD06655161
  • Cas Number : 857041-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10826
Availability details for
Quantity
  • Availability
  • Add to Cart