ENA040899092 Enamine

4-pentyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06655164
  • Cas Number : 27106-02-1
  • Molecular Weight : 171.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-10831

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10831
Availability details for
Quantity
  • Availability
  • Add to Cart