ENA040899098 Enamine

N-[4-(chloromethyl)-1,3-thiazol-2-yl]-N-(2,3-dimethylphenyl)acetamide

  • MDL number MFCD06655169
  • Cas Number : 857041-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10839

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10839
Availability details for
Quantity
  • Availability
  • Add to Cart