ENA040899107 Enamine

4-methoxy-3-(methylsulfamoyl)benzoic acid

  • MDL number MFCD06355954
  • Cas Number : 576169-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10852
Availability details for
Quantity
  • Availability
  • Add to Cart