ENA040899108 Enamine

4-methoxy-3-sulfamoylbenzoic acid

  • MDL number MFCD06355949
  • Cas Number : 20532-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10853
Availability details for
Quantity
  • Availability
  • Add to Cart