ENA040899111 Enamine

2-fluoro-5-(methylsulfamoyl)benzoic acid

  • MDL number MFCD06355951
  • Cas Number : 716358-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10860
Availability details for
Quantity
  • Availability
  • Add to Cart