ENA040899113 Enamine

5-(dimethylsulfamoyl)-2-fluorobenzoic acid

  • MDL number MFCD06355944
  • Cas Number : 91103-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10862
Availability details for
Quantity
  • Availability
  • Add to Cart