ENA040899114 Enamine

3,4-dihydro-2H-1,5-benzodioxepine-7-thiol

  • MDL number MFCD06355957
  • Cas Number : 786728-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10866
Availability details for
Quantity
  • Availability
  • Add to Cart