ENA040899118 Enamine

5-(piperazin-1-yl)thiophene-2-carbaldehyde

  • MDL number MFCD06375953
  • Cas Number : 851208-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-69
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10879
Availability details for
Quantity
  • Availability
  • Add to Cart