ENA040899119 Enamine

2-chloro-1-[4-(2,3-dihydro-1,4-benzodioxine-2-carbonyl)piperazin-1-yl]ethan-1-one

  • MDL number MFCD06655173
  • Cas Number : 857041-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10880
Availability details for
Quantity
  • Availability
  • Add to Cart