ENA040899120 Enamine

4-(1-aminoethyl)-N,N-diethylbenzene-1-sulfonamide

  • MDL number MFCD06655174
  • Cas Number : 854035-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-114
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10881
Availability details for
Quantity
  • Availability
  • Add to Cart