ENA040899123 Enamine

2-[5-(diethylsulfamoyl)-2-oxo-1,2-dihydropyridin-1-yl]acetic acid

  • MDL number MFCD06655178
  • Cas Number : 854035-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-189
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10885
Availability details for
Quantity
  • Availability
  • Add to Cart