ENA040899124 Enamine

2-(4-cyanophenoxy)propanoic acid

  • MDL number MFCD06655179
  • Cas Number : 118618-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-159
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10887
Availability details for
Quantity
  • Availability
  • Add to Cart