ENA040899126 Enamine

2-[2-oxo-2-(pyrrolidin-1-yl)ethoxy]benzaldehyde

  • MDL number MFCD06655181
  • Cas Number : 879362-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-86
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-10892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10892
Availability details for
Quantity
  • Availability
  • Add to Cart