ENA040899132 Enamine

N-[4-(chloromethyl)-1,3-thiazol-2-yl]-N-(2,4-difluorophenyl)acetamide

  • MDL number MFCD06655185
  • Cas Number : 854035-85-1
  • Molecular Weight : 302.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-10907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10907
Availability details for
Quantity
  • Availability
  • Add to Cart