ENA040899134 Enamine

N-(3-chlorophenyl)-N-(4-formyl-1,3-thiazol-2-yl)acetamide

  • MDL number MFCD06655187
  • Cas Number : 854035-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10909

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10909
Availability details for
Quantity
  • Availability
  • Add to Cart