ENA040899138 Enamine

N-(2-ethylphenyl)-N-[4-(isothiocyanatomethyl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD06655191
  • Cas Number : 879362-37-5
  • Molecular Weight : 317.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-97
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10913
Availability details for
Quantity
  • Availability
  • Add to Cart