ENA040899139 Enamine

N-(2,4-dimethylphenyl)-N-[4-(isothiocyanatomethyl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD06655192
  • Cas Number : 885460-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10914

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10914
Availability details for
Quantity
  • Availability
  • Add to Cart