ENA040899142 Enamine

4-(3-methylbutyl)-5-(phenoxymethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06655195
  • Cas Number : 854035-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10918

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10918
Availability details for
Quantity
  • Availability
  • Add to Cart