ENA040899146 Enamine

N-[4-(chloromethyl)-1,3-thiazol-2-yl]-N-(2,6-dimethylphenyl)acetamide

  • MDL number MFCD06655200
  • Cas Number : 854035-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10923

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10923
Availability details for
Quantity
  • Availability
  • Add to Cart