ENA040899151 Enamine

4-methoxy-3-[(2-methoxyethyl)sulfamoyl]benzoic acid

  • MDL number MFCD06349003
  • Cas Number : 299181-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-10562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10562
Availability details for
Quantity
  • Availability
  • Add to Cart