ENA040899153 Enamine

2-chloro-5-[(isopropylamino)sulfonyl]benzoic acid

  • MDL number MFCD05988614
  • Cas Number : 74138-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...149
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10564

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10564
Availability details for
Quantity
  • Availability
  • Add to Cart