ENA040899160 Enamine

2-chloro-1-[2,5-dimethyl-1-(1-phenylethyl)-1H-pyrrol-3-yl]ethan-1-one

  • MDL number MFCD06349000
  • Cas Number : 784172-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10571
Availability details for
Quantity
  • Availability
  • Add to Cart