ENA040899161 Enamine

2-methyl-5-[(propan-2-yl)sulfamoyl]benzoic acid

  • MDL number MFCD05988747
  • Cas Number : 701272-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10572
Availability details for
Quantity
  • Availability
  • Add to Cart