ENA040899162 Enamine

2-mercapto-3-pyridin-3-ylquinazolin-4(3H)-one

  • MDL number MFCD00113288
  • Cas Number : 119426-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10710

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10710
Availability details for
Quantity
  • Availability
  • Add to Cart