ENA040899164 Enamine

N-(3,4-dimethylphenyl)-N-(4-formyl-1,3-thiazol-2-yl)acetamide

  • MDL number MFCD06347914
  • Cas Number : 771499-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10503

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10503
Availability details for
Quantity
  • Availability
  • Add to Cart