ENA040899166 Enamine

4-(2,5-dimethylphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03900781
  • Cas Number : 669748-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-10576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10576
Availability details for
Quantity
  • Availability
  • Add to Cart