ENA040899167 Enamine

N-(2-fluorophenyl)-N-(4-formyl-1,3-thiazol-2-yl)acetamide

  • MDL number MFCD06347915
  • Cas Number : 771499-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10504
Availability details for
Quantity
  • Availability
  • Add to Cart