ENA040899175 Enamine

7-chloro-3-(2-isopropylphenyl)-2-mercaptoquinazolin-4(3H)-one

  • MDL number MFCD06347919
  • Cas Number : 771499-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10506

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10506
Availability details for
Quantity
  • Availability
  • Add to Cart