ENA040899186 Enamine

2-{[1-(2-ethoxyphenyl)-1H-1,2,3,4-tetrazol-5-yl]sulfanyl}acetic acid

  • MDL number MFCD04035349
  • Cas Number : 436095-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-127
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10588

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10588
Availability details for
Quantity
  • Availability
  • Add to Cart