ENA040899200 Enamine

2-chloro-N-(1,1-dioxo-1lambda6-thiolan-3-yl)-N-phenylacetamide

  • MDL number MFCD03038715
  • Cas Number : 507458-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10599
Availability details for
Quantity
  • Availability
  • Add to Cart