ENA040899205 Enamine

{[(2E)-3-phenylprop-2-enoyl]amino}acetic acid

  • MDL number MFCD00136326
  • Cas Number : 16534-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10753

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10753
Availability details for
Quantity
  • Availability
  • Add to Cart