ENA040899214 Enamine

2-phenyl-2-(pyrrolidin-1-yl)ethan-1-amine

  • MDL number MFCD01631939
  • Cas Number : 31466-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10762
Availability details for
Quantity
  • Availability
  • Add to Cart