ENA040899215 Enamine

2-(pyrrolidin-1-yl)-2-(thiophen-2-yl)ethan-1-amine

  • MDL number MFCD06655137
  • Cas Number : 142919-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10763
Availability details for
Quantity
  • Availability
  • Add to Cart