ENA040899216 Enamine

2-(N-methyl5-chlorothiophene-2-sulfonamido)acetic acid

  • MDL number MFCD06335631
  • Cas Number : 749897-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10522
Availability details for
Quantity
  • Availability
  • Add to Cart