ENA040899225 Enamine

4-chloro-2-(4-acetamidobenzenesulfonamido)benzoic acid

  • MDL number MFCD06655062
  • Cas Number : 851109-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-10524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10524
Availability details for
Quantity
  • Availability
  • Add to Cart