ENA040899228 Enamine

1-[3,5-bis(thiophen-2-yl)-4,5-dihydro-1H-pyrazol-1-yl]-2-chloroethan-1-one

  • MDL number MFCD03685634
  • Cas Number : 799837-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10776
Availability details for
Quantity
  • Availability
  • Add to Cart