ENA040899229 Enamine

5-(pyrrolidine-1-sulfonyl)-1,2-dihydropyridin-2-one

  • MDL number MFCD07313523
  • Cas Number : 627843-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10777
Availability details for
Quantity
  • Availability
  • Add to Cart