ENA040899234 Enamine

4-chloro-3-{[(2,4-dichlorobenzyl)amino]sulfonyl}benzoic acid

  • MDL number MFCD06347469
  • Cas Number : 782462-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10782

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10782
Availability details for
Quantity
  • Availability
  • Add to Cart