ENA040899240 Enamine

4-(bromomethyl)-N-phenylbenzamide

  • MDL number MFCD06655146
  • Cas Number : 566885-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10788

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10788
Availability details for
Quantity
  • Availability
  • Add to Cart