ENA040899246 Enamine

2-methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid

  • MDL number MFCD03001420
  • Cas Number : 272437-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10623
Availability details for
Quantity
  • Availability
  • Add to Cart