ENA040899247 Enamine

4-(3,4-dimethylphenyl)-5-pyridin-4-yl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD05134116
  • Cas Number : 117080-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260...262
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10795

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10795
Availability details for
Quantity
  • Availability
  • Add to Cart